Learn More
Kemtec™ Anti-Rh, Anti-D Serum Set (5mL)
Catalog No. S06448
Change view
Click to view available options
Grade:
8 to 14
Catalog No. Grade
S06448 8 to 14
1 options
Choking hazard (1) Small parts. Not for children under 3 yrs.
Catalog No. S06448 Supplier Customer Channels Group (Ccg) Supplier No. 3246
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Choking hazard (1) Small parts. Not for children under 3 yrs.
Highly specific monoclonal antiserum to Rh factor tested for HbsAg and HTLV-III (AIDS).
Standard laboratory practices should be used and all biological products should be treated as if capable of transmitting disease. Product is perishable and requires special shipping. Refrigerate to prolong shelf life.
Kit Contains:
  • Anti-D serum
  • Dropper
  • Approximately 36 tests
  • For grades 9 to 14
Specifications
Grade 8 to 14
Includes Anti-d serum, dropper
Product Type Anti-Rh, Anti-D Serum Set