Learn More
Kemtec™: Analysis of Fingerprints
Encourages students to learn and experiment with dusting and developing latent prints.
Supplier:  Kemtec KMT10504
Catalog No. S25716
Only null left
Includes: Brushes, metal spoons, slides, cellophane tape, magnifying glasses, jars with caps, aluminum foil, ink pads, tweezers, set of FBI fingerprint classifications, gloves, filter paper, index cards, super glue, chemicals, 22 student data sheets, instructions
Requires : Fume hood
Description
Description
  • Define latent fingerprints found on various surfaces
  • Compare prints using standards developed by the FBI to identify a would-be suspect
  • Prepare evidence for analysis and identification of fingerprints
  • Develop prints on paper using ninhydrin and Lugol's iodine method
  • Visualize fingerprint on metal spoon with cyanoacrylate fuming method
  • Grades 7-14
  • Class size: 24 students (six groups of four)
Specifications
Specifications
National standards
Fingerprint analysis
Fume hood, (6) 1 qt. jars or 1L beakers
Analysis of Fingerprints Kit
7 to 14
Instructor manual, student data sheets, msds, “fbi” fingerprint classification sheets, bond paper, filter paper, gloves, pipet, fingerprint brushes, magnifying glasses, forceps, jars with caps, metal spoons, aluminum foil, superglue, ink pads, scotch tape
24 Students
Product Suggestions
Product Suggestions