missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Kemptide

For cellular and molecular biology applications

$259.49 - $259.49

Specifications

Purification Method HPLC
Content And Storage -30°C to -10°C
Format Powder
Molecular Weight (g/mol) 771.91
Product Type Kemptide
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ60591LB0
View Documents
Thermo Scientific Chemicals
J60591LB0
5 mg
Each for $259.49
Only null left
 
Description

Description

Kemptide, phosphate acceptor, is shown to be a basic serine-containing heptapeptide corresponding to a sequence from pig liver pyruvate kinase. This synthetic peptide is a substrate of PKA (cyclic AMP-dependent protein kinase) and is phosphorylated and activated after microinjection into Xenopus oocytes. Exposure of Kemptide to insulin has demonstrated an increase in activation of this agent, which can be inhibited by the catalytic subunit of phosphatase 2A.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

HPLC
Powder
Kemptide
Leu-Arg-Arg-Ala-Ser-Leu-Gly
-30°C to -10°C
771.91
RUO - research use only
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only