Learn More
Chemglass Inc Clamp, Keck, Blue, Standard Taper, Fits Joint Sizes 19/22 & 19/38
Supplier: Chemglass Inc CG14504
Clamp, Keck, Blue, Standard Taper, Fits Joint Sizes 19/22 & 19/38. Made of Delrin, the clamp is non-corrosive and will not scratch glass. Can be used to 140 C. Color coded for easy identification. SOLD IN PACKAGES OF 10.