Learn More
KD Scientific Stainless-Steel Syringes
Durable, electron-beam construction withstands high force, high pressure applications
Supplier:  KD Scientific 780814
 View more versions of this product
Catalog No. 1493148
Only null left
Description
Description
Eliminates hazards of glass syringe breakage.
  • Compatible with most syringe pumps
  • Fully autoclavable for reuse
  • Adaptable to Luer Lock or Swagelok™ fittings
  • Resistant to most chemicals
Specifications
Chemical Identifiers
Autoclavable
Stainless Steel
Syringe Pump
Specifications
Autoclavable
1-37/100 in.
34.7 mm
Swagelok 1/4 in. Syringe
100 mL
Stainless Steel
1-1/5 in.
38.1 mm
1500 psi
Syringe Pump
Product Suggestions
Product Suggestions