Learn More
KD Scientific Stainless-Steel Syringes
Durable, electron-beam construction withstands high force, high pressure applications
Supplier: KD Scientific 780810
Description
Eliminates hazards of glass syringe breakage.
- Compatible with most syringe pumps
- Fully autoclavable for reuse
- Adaptable to Luer Lock or Swagelok™ fittings
- Resistant to most chemicals
Chemical Identifiers
| Autoclavable | |
| Stainless Steel | |
| Syringe Pump |
Specifications
| Autoclavable | |
| 1-37/100 in. | |
| 34.7 mm | |
| Swagelok 1/8 in. Syringe | |
| 100 mL |
| Stainless Steel | |
| 1-1/5 in. | |
| 38.1 mm | |
| 1500 psi | |
| Syringe Pump |