Learn More
SignalChem Biotech EZH2 (Y641S) Protein 1000ug
Catalog No. 502574970
Encompass
Recombinant full-length human EZH2 (Y641S) protein was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
Catalog No. 502574970 Supplier SignalChem Biotech Supplier No. E39632FG1000
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Recombinant full-length human EZH2 (Y641S) protein was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.