Learn More
Thermo Scientific Chemicals Kartogenin
$307.96 - $307.96
Chemical Identifiers
| CAS | 4727-31-5 |
|---|---|
| Molecular Formula | C20H15NO3 |
| Molecular Weight (g/mol) | 317.34 |
| InChI Key | SLUINPGXGFUMLL-UHFFFAOYSA-N |
| IUPAC Name | 2-({[1,1'-biphenyl]-4-yl}carbamoyl)benzoic acid |
| SMILES | OC(=O)C1=CC=CC=C1C(=O)NC1=CC=C(C=C1)C1=CC=CC=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC464662500
|
Thermo Scientific Chemicals
464662500 |
250 mg |
Each for $307.96
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 4727-31-5 | |
| 317.34 | |
| 2-({[1,1'-biphenyl]-4-yl}carbamoyl)benzoic acid |
| C20H15NO3 | |
| SLUINPGXGFUMLL-UHFFFAOYSA-N | |
| OC(=O)C1=CC=CC=C1C(=O)NC1=CC=C(C=C1)C1=CC=CC=C1 |
Specifications
| 4727-31-5 | |
| C20H15NO3 | |
| Solubility aq. soln.: .. Other solubilities: slightly soluble in dmso and methanol | |
| OC(=O)C1=CC=CC=C1C(=O)NC1=CC=C(C=C1)C1=CC=CC=C1 | |
| 317.34 | |
| Kartogenin |
| >280.0°C | |
| 250 mg | |
| SLUINPGXGFUMLL-UHFFFAOYSA-N | |
| 2-({[1,1'-biphenyl]-4-yl}carbamoyl)benzoic acid | |
| 317.34 |
Safety and Handling
GHS H Statement Harmful if swallowed.
Causes serious eye irritation.
Very toxic to aquatic life with long lasting effects.
GHS P Statement IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wash face, hands and any exposed skin thoroughly after handling.
Wear eye protection/face protection.
If eye irritation persists: Get medical advice/attention.
GHS Signal Word: Warning
RUO – Research Use Only