missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Kartogenin

Catalog No. AC464662500
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC464662500 250 mg
AC464660010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC464662500 Supplier Thermo Scientific Chemicals Supplier No. 464662500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 4727-31-5 | C20H15NO3 | 317.34 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4727-31-5
Molecular Formula C20H15NO3
Molecular Weight (g/mol) 317.34
InChI Key SLUINPGXGFUMLL-UHFFFAOYSA-N
IUPAC Name 2-({[1,1'-biphenyl]-4-yl}carbamoyl)benzoic acid
SMILES OC(=O)C1=CC=CC=C1C(=O)NC1=CC=C(C=C1)C1=CC=CC=C1

Specifications

Melting Point >280.0°C
Quantity 250 mg
Solubility Information Solubility aq. soln.: .. Other solubilities: slightly soluble in dmso and methanol
Formula Weight 317.34
Chemical Name or Material Kartogenin
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Harmful if swallowed.

Causes serious eye irritation.

Very toxic to aquatic life with long lasting effects.

Precautionary Statement GHS P Statement IF SWALLOWED: rinse mouth.

Do NOT induce vomiting.

Call a POISON CENTER or doctor/physician if you feel unwell.

Wash face, hands and any exposed skin thoroughly after handling.

Wear eye protection/face protection.

If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only