missing translation for 'onlineSavingsMsg'
Learn More

Karaya Gum

Catalog No. AAJ6184436
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6184436 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ6184436 Supplier Thermo Scientific Chemicals Supplier No. J6184436

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 9000-36-6

Used as a thickener and emulsifier

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 9000-36-6
MDL Number MFCD00131251
Synonym Gum karaya from sterculia tree; Tragacanth Indian

Specifications

Color Brown to Yellow
Quantity 500 g
Merck Index 14,5284
Physical Form Powder
Chemical Name or Material Karaya Gum
EINECSNumber 232-539-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only