Learn More
Karaya Gum, FCC, Spectrum™ Chemical
KA115, 9000-36-6
$356.17 - $11115.87
Chemical Identifiers
CAS 9000-36-6
View More Specs
Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18603628
View Documents
Spectrum Chemical Mfg Cor
KA115500GM
500 g Poly Bottle
Each for $356.17
Only null left
18603629
View Documents
Spectrum Chemical Mfg Cor
KA11525KG
2.5 kg Poly Pail
Each for $1,275.50
Only null left
18603630
View Documents
Spectrum Chemical Mfg Cor
KA11545KGBL
45 kg Fiber Drum N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
Spectrum™ Chemical Karaya Gum, FCC , a vegetable gum, is used as a thickener and emulsifier in foods. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
Chemical Identifiers
9000-36-6
Specifications
9000-36-6
1%
FCC
20%
100%
500 g
3%
Poly Bottle
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'