missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kappler™ Zytron 200 Totally Encapsulating Suits
Supplier: Kappler™ Z2H371SMMD
Specifications
| Medium | |
| Gray | |
| Sealed, Taped | |
| Flat | |
| Ideally used with wider range of chemicals | |
| 5.4 kg |
| Zytron™ | |
| Double Storm Flap with Hook and Loop Seal | |
| Chemicals | |
| Flat back, rear entry, side air inlet, double storm flap with hook and loop closure, cloth zipper | |
| 12 lb. |