Learn More
Kappler™ Zytron™ 100 Chemical-Resistant Coveralls
Provides basic chemical resistance
Supplier:  Kappler™ Z1S414XPSMMD
 View more versions of this product
Catalog No. 19-167-841B
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Explore available promotions
Description
Description
  • Zippered front
  • Seam type available in bound and serged
  • Available with attached hood and/or boots and/or elastic wrists and ankles
  • Ideal for applications where workers need protection from dry particulates and light splash
Compliance
Meets ASTM™ F739 industrial chemical testing requirements.
Specifications
Specifications
Medium
Yellow
Elastic
Front Zipper
Chemicals
Attached hood and boots, elastic wrists
Ideal for applications where workers need protection from dry particulates and light splash
6.4 kg
Zytron™
Elastic, Attached Boots
Hooded
Serged
ASTM D751, D3787
Attached hood and boots, elastic wrists
14 lb.
Product Suggestions
Product Suggestions