Learn More
Kappler™ Zytron™ 100 Chemical-Resistant Coveralls
Provides basic chemical resistance
Supplier:  Kappler™ Z1B428XPLGXL
 View more versions of this product
Catalog No. 19-167-840C
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Explore available promotions
Description
Description
  • Zippered front
  • Seam type available in bound and serged
  • Available with attached hood and/or boots and/or elastic wrists and ankles
  • Ideal for applications where workers need protection from dry particulates and light splash
Compliance
Meets ASTM™ F739 industrial chemical testing requirements.
Specifications
Specifications
Large
Yellow
Elastic
Front Zipper
Chemicals
Attached hood, elastic wrists and ankles
Ideal for applications where workers need protection from dry particulates and light splash
5.4 kg
Zytron™
Elastic
Hooded
Bound
ASTM D751, D3787
Attached hood, elastic wrists and ankles
12 lb.
Product Suggestions
Product Suggestions