Learn More
Kappler™ Zytron™ 100 Chemical-Resistant Coveralls
Provides basic chemical resistance
Supplier:  Kappler™ Z1S428XP2X3X
 View more versions of this product
Catalog No. 19-167-839E
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Explore available promotions
Description
Description
  • Zippered front
  • Seam type available in bound and serged
  • Available with attached hood and/or boots and/or elastic wrists and ankles
  • Ideal for applications where workers need protection from dry particulates and light splash
Compliance
Meets ASTM™ F739 industrial chemical testing requirements.
Specifications
Specifications
2X-Large
Yellow
Elastic
Front Zipper
Chemicals
Attached hood, elastic wrists and ankles
Ideal for applications where workers need protection from dry particulates and light splash
5.4 kg
Zytron™
Elastic
Hooded
Serged
ASTM D751, D3787
Attached hood, elastic wrists and ankles
12 lb.
Product Suggestions
Product Suggestions