missing translation for 'onlineSavingsMsg'
Learn More

Kappler™ Zytron™ 100 Chemical-Resistant Coveralls

Catalog No. 19167838C
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
Seam Type:
Bound
Serged
Includes:
Attached hood and boots, elastic wrists
Attached hood, elastic wrists and ankles
Collar, elastic wrists and ankles
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Seam Type Includes
19167838C Large Bound Collar, elastic wrists and ankles
19167838B Medium Bound Collar, elastic wrists and ankles
19167838E 2X-Large Bound Collar, elastic wrists and ankles
19167839E 2X-Large Serged Attached hood, elastic wrists and ankles
19167840C Large Bound Attached hood, elastic wrists and ankles
19167840B Medium Bound Attached hood, elastic wrists and ankles
19167840D X-Large Bound Attached hood, elastic wrists and ankles
19167840E 2X-Large Bound Attached hood, elastic wrists and ankles
19167841B Medium Serged Attached hood and boots, elastic wrists
19167841D X-Large Serged Attached hood and boots, elastic wrists
19167842C Large Bound Attached hood and boots, elastic wrists
19167842B Medium Bound Attached hood and boots, elastic wrists
19167842D X-Large Bound Attached hood and boots, elastic wrists
19167842E 2X-Large Bound Attached hood and boots, elastic wrists
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
Catalog No. 19167838C Supplier Kappler™ Supplier No. Z1B417XPLGXL
Only null left

Provides basic chemical resistance

  • Zippered front
  • Seam type available in bound and serged
  • Available with attached hood and/or boots and/or elastic wrists and ankles
  • Ideal for applications where workers need protection from dry particulates and light splash

Compliance

Meets ASTM™ F739 industrial chemical testing requirements.

Specifications

Size Large
Material Zytron™
Color Yellow
Ankle Style Elastic
Cuff Style Elastic
Closure Type Front Zipper
Seam Type Bound
Resistance Chemicals
Certifications/Compliance ASTM D751, D3787
Description Collar, elastic wrists and ankles
Includes Collar, elastic wrists and ankles
For Use With (Application) Ideal for applications where workers need protection from dry particulates and light splash
Weight (English) 11 lb.
Weight (Metric) 5 kg
Show More Show Less