missing translation for 'onlineSavingsMsg'
Learn More

Kappler™ ProVent™ 7000 Isolation Gowns, Tie Closures Neck and Waist

Catalog No. 19999344C
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Cuff Style:
Elastic
Knit
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Cuff Style
19999344C Medium Elastic
19999344A Small Knit
19999344B Large Knit
19999344E X-Large Knit
19999344F 2X-Large Knit
18025653 3X-Large Elastic
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 19999344C Supplier Kappler™ Supplier No. PVS112WHMD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Breathable, microporous fabric gowns offer general hazard protection for a variety of work environments from maintenance to general healthcare. Choose from elastic wrist or knit cuff options.

Lightweight composite fabric utilizes microporous film technology to provides protection against hazardous particles and light liquid splash and maintain strength wet or dry

  • Wraparound style opens to back
  • Knit cuffs or elastic wrists
  • Two neck ties, two waist ties
  • Nonsterile
  • White

Order Info

  • Front length: small 38.75 in. (98.4 cm), medium 39.75 in. (101 cm), large 40.75 in. (103.5 cm). Back length of all sizes: 3 in. (7.6 cm) longer than front

Compliance

Meets ASTM™ F1670 and ASTM F1671 standards.

Specifications

Size Medium
For Use With (Application) Maintenance, Spray Painting, Healthcare, Clinical
Gender Unisex
No. of Pockets None
Product Type General Purpose Gown
Style Wrap-around
Material Microporous Coated Fabric
Color White
Closure Type Neck and Waist Ties
Disposable Single-use
Sterility Non-sterile
Cuff Style Elastic
Model PVS112WHMD
Seam Type Serged
Sleeve Style Long Sleeve
Certifications/Compliance ASTM F1670, ASTM F1671
Weight (English) 17 lbs.
Autoclavable Not Autoclavable
Show More Show Less