missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kappler™ ProVent™ 7000 Isolation Gowns, Tie Closures Neck and Waist
Description
Lightweight composite fabric utilizes microporous film technology to provides protection against hazardous particles and light liquid splash and maintain strength wet or dry
- Wraparound style opens to back
- Knit cuffs or elastic wrists
- Two neck ties, two waist ties
- Nonsterile
- White
Order Info
- Front length: small 38.75 in. (98.4 cm), medium 39.75 in. (101 cm), large 40.75 in. (103.5 cm). Back length of all sizes: 3 in. (7.6 cm) longer than front
Compliance
Meets ASTM™ F1670 and ASTM F1671 standards.
Specifications
Specifications
| Style | Wrap-around |
| Size | Small |
| No. of Pockets | None |
| For Use With (Application) | Maintenance, Spray Painting, Healthcare, Clinical |
| Gender | Unisex |
| Product Type | General Purpose Gown |
| Material | Microporous Coated Fabric |
| Color | White |
| Closure Type | Neck and Waist Ties |
| Disposable | Single-use |
| Show More |