missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kappler™ ProVent™ 10000 Wrap-Around Gowns
ProVent 10,000 wrap-around gowns feature a patented microporous film composite fabric that offers a wide range of barrier protection.
Supplier: Kappler™ PPN101BU3X
Description
- ProVent 10,000 is a patented microporous film composite fabric that offers a wide range of barrier protection against blood-borne pathogens, body fluids, and viral threats
- It passes both ASTM F1670 blood penetration and ASTM F1671 viral penetration tests
- High moisture vapor transmission rate (MVTR) provides a more comfortable garment ideal for biohazard work applications
Compliance
Meets ASTM™ F1670 and ASTM F1671 standards.
Specifications
| General Purpose Gown | |
| None | |
| 3X-Large | |
| Blue | |
| Single-use | |
| Knit | |
| Ultrasonic | |
| ASTM F1670, ASTM F1671 |
| Biohazard Cleanup, Healthcare, Maintenance | |
| Unisex | |
| Composite Fabric | |
| Hook and Loop | |
| Non-sterile | |
| MN101BU3X | |
| Long Sleeve | |
| Not Autoclavable |