missing translation for 'onlineSavingsMsg'
Learn More

Kappler™ ProVent™ 10000 Wrap-Around Gowns

Catalog No. 17988497G
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17988497G 4X-Large
17988497A Small
17988497C Large
17988497B Medium
17988497D X-Large
17988497E 2X-Large
17988497F 3X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 17988497G Supplier Kappler™ Supplier No. PPN101BU4X

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

ProVent 10,000 wrap-around gowns feature a patented microporous film composite fabric that offers a wide range of barrier protection.

  • ProVent 10,000 is a patented microporous film composite fabric that offers a wide range of barrier protection against blood-borne pathogens, body fluids, and viral threats
  • It passes both ASTM F1670 blood penetration and ASTM F1671 viral penetration tests
  • High moisture vapor transmission rate (MVTR) provides a more comfortable garment ideal for biohazard work applications

Compliance

Meets ASTM™ F1670 and ASTM F1671 standards.

Specifications

Size 4X-Large
For Use With (Application) Biohazard Cleanup, Healthcare, Maintenance
Gender Unisex
No. of Pockets None
Product Type General Purpose Gown
Material Composite Fabric
Color Blue
Closure Type Hook and Loop
Disposable Single-use
Sterility Non-sterile
Cuff Style Knit
Model MN101BU4X
Seam Type Ultrasonic
Sleeve Style Long Sleeve
Certifications/Compliance ASTM F1670, ASTM F1671
Autoclavable Not Autoclavable
Show More Show Less