Learn More
Kappler™ ProVent™ Plus Medical Boot Covers
Catalog No. 19315325
Change view
missing translation for 'orderingAttributeHoverText'
:
Size: Large
Catalog No.
19315325 Size: Large
1 options
Catalog No. 19315325
Supplier: Kappler™ PPN026BULG
Only null left
Non-sterile medical boot covers offer a wide range of barrier protection against blood, body fluids, and viral threats.
  • Knee-high boot covers with hook and loop closure
  • Elastic at top half of boot, 2-ply sole
  • Constructed from a patented microporous film composite
  • Wide range of barrier protection against blood, body fluids and viral threats
  • Passes both ASTM F1670 blood penetration and ASTM 1671 viral penetration tests
  • High moisture vapor transmission rate (MVTR) results in a cool, comfortable garment
  • Ideal for biohazard cleanup
  • Non-sterile
Specifications
Size Large
Color Blue
Closure Type Elastic with Hook and Loop Ankle
Certifications/Compliance ASTM 1670, ASTM 1671
Product Type Boot Covers