missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Kallikrein Inhibitor
$377.78
Chemical Identifiers
| CAS | 97145-43-2 |
|---|---|
| Molecular Formula | C35H55N11O9 |
| Synonym | N-Acetyl-Pro-Phe-Arg-Ser-Val-Gln-NH{2}; KKI 5 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
AAJ66234LB0
|
Thermo Scientific Chemicals
J66234LB0 |
5 mg |
Each for $377.78
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 97145-43-2 | |
| C35H55N11O9 | |
| N-Acetyl-Pro-Phe-Arg-Ser-Val-Gln-NH{2}; KKI 5 |
Specifications
| White to off-white | |
| 97145-43-2 | |
| 773.89 | |
| Soluble in water at 1mg/ml | |
| Solid | |
| CC(C)[C@H](NC(=O)[C@H](CO)NC(=O)[C@H](CCCN=C(N)N)NC(=O)[C@H](CC1=CC=CC=C1)NC(=O)[C@@H]1CCCN1C(C)=O)C(=O)N[C@@H](CCC(N)=O)C(N)=O | |
| 773.89 |
| -30°C to -10°C | |
| C35H55N11O9 | |
| N-Acetyl-Pro-Phe-Arg-Ser-Val-Gln-NH{2}; KKI 5 | |
| Kallikrein Inhibitor | |
| OOZYLOMMGMLDHT-BIVGDOEESA-N | |
| (2S)-2-[(2S)-2-[(2S)-2-[(2S)-2-[(2S)-2-{[(2S)-1-acetylpyrrolidin-2-yl]formamido}-3-phenylpropanamido]-5-[(diaminomethylidene)amino]pentanamido]-3-hydroxypropanamido]-3-methylbutanamido]pentanediamide |
Safety and Handling
Recommended Storage : Store at -20°C
RUO – Research Use Only