missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Kainic acid monohydrate, 96%

Catalog No. AAJ66590MA
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ66590MA 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ66590MA Supplier Thermo Scientific Chemicals Supplier No. J66590MA

CAS: 58002-62-3 | C10H17NO5, C10H15NO4 | 231.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 58002-62-3
Molecular Formula C10H17NO5
Molecular Weight (g/mol) 231.25
InChI Key FZNZRJRSYLQHLT-UHFFFAOYNA-N
Synonym 2-Carboxy-3-carboxymethyl-4-isopropenylpyrrolidine; (-)-(α)-Kainic Acid
IUPAC Name 3-(carboxymethyl)-4-(prop-1-en-2-yl)pyrrolidine-2-carboxylic acid hydrate
SMILES O.CC(=C)C1CNC(C1CC(O)=O)C(O)=O

Specifications

Quantity 10 mg
Description White to pale yellow
Content And Storage 2°C to 8°C
Formula Weight 213.23
Sensitivity Light Sensitive
Melting Point 251°C
Product Type Kainic Acid Monohydrate, 98%
Form Crystalline solid
Assay 98%
Recommended Storage Keep cold

RUO – Research Use Only