missing translation for 'onlineSavingsMsg'
Learn More

JWH-073 4-Butanoic Acid Metabolite Solution, 100 μg/mL in Methanol, Ampule of 1 mL, Certified Reference Material, Cerilliant™

Catalog No. 01001951
Change view
Click to view available options
Grade:
Certified
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
01001951 Certified
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01001951 Supplier Sartorius Corporation Supplier No. PMP01P000011
Only null left
  • A secondary urinary metabolite of JWH-073, a synthetic cannabinoid found in several versions of the herbal mixture Spice.
  • Our certified Spice cannabinoid solution standards are DEA-exempt allowing laboratories to order these standards without DEA paperwork.

Specifications

Chemical Name or Material JWH-073 4-Butanoic acid Metabolite
CAS 1307803-52-6
Packaging Ampule
Molecular Formula C23H19NO3
Molecular Weight (g/mol) 357.4
Grade Certified
Concentration or Composition (by Analyte or Components) 100 μg/mL
For Use With (Equipment) Liquid Chromatography (LC); Gas Chromatography (GC)
For Use With (Application) Forensics and Toxicology