missing translation for 'onlineSavingsMsg'
Learn More
Learn More
JWH-018 5-Pentanoic Acid Metabolite Solution, 100 μg/mL in Methanol, Ampule of 1 mL, Certified Reference Material, Cerilliant™
Description
- A secondary urinary metabolite of JWH-018, a synthetic cannabinoid found in many blends of the herbal mixture Spice also known as K2, Genie, or Demon.
- Our certified Spice cannabinoid solution standards are DEA-exempt allowing laboratories to order these standards without DEA paperwork.
Specifications
Specifications
| Chemical Name or Material | JWH-018 5-Pentanoic acid Metabolite |
| CAS | 1254475-87-0 |
| Molecular Formula | C24H21NO3 |
| Concentration | 100 μg/mL |
| Molecular Weight (g/mol) | 371.43 |
| Grade | Certified |
| Packaging | Ampule |