missing translation for 'onlineSavingsMsg'
Learn More

Justrite™ Tip-Over Protection System for Drum Funnel

Catalog No. 2870594
Change view
Click to view available options
Material:
Brass
Galvanized Steel
Product Type:
Drum Funnel
Tip-over Protection Kit
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material Product Type
2870594 Brass Tip-over Protection Kit
2870593 Galvanized Steel Drum Funnel
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 2870594 Supplier Justrite™ Supplier No. 08214

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

Model 08214 (Tip-Over Protection Kit Only)

  • Self-close valve with 5.1cm (2 in.) bung adapter
  • Brass vent with 1.9cm (0.75 in.) bung adapter

Model 08203 (Funnel with Tip-Over Protection Kit)

  • Funnel with self-closing ball valve and vent adapter
  • 15.2cm (6 in.) flame arrester
  • Tip-over protection kit

Secure protection for 5.1cm (2 in. ) drum bung

  • Funnel made of galvanized steel
  • Vent made of brass
  • Self-closing valve and cover

Certifications

FM approved

Specifications

Certifications/Compliance FM
Product Type Tip-over Protection Kit
Description Self Close
Material Brass
Includes Self-close valve with 5.1cm bung adapter and brass vent with 1.9cm bung adapter