missing translation for 'onlineSavingsMsg'
Learn More

Justrite™ Sure-Grip™ EX Hazardous Material Steel Safety Cabinet
GREENER_CHOICE

Keep storage of your hazardous materials compliant with the Justrite™ Sure-Grip™ EX Hazmat Safety Cabinet.

$5,401.45 - $6,718.96

Specifications

Color Blue
Product Type Combi Safety Storage Cabinet
Door Style Self-close
For Use With (Application) Hazardous materials
No. of Doors 2
View More Specs

Products 2
Catalog Number Mfr. No. Load Bearing Capacity Dimensions (D x W x H) Price Quantity & Availability  
Catalog Number Mfr. No. Load Bearing Capacity Dimensions (D x W x H) Price Quantity & Availability  
19414801
GREENER_CHOICE
View Documents Promotion Details
Justrite™
869028
90 gal. 34 x 43 x 65 in.
Each for $6,718.96
Only null left
 
19414800
GREENER_CHOICE
View Documents Promotion Details
Justrite™
866028
60 gal. 34 x 34 x 65 in.
Each for $5,401.45
Only null left
 
Description

Description

  • Double-wall construction, dual vents, grounding wire connections, adjustable shelves
  • Leakproof sills, three-point self-latching doors and leveling feet
  • Self-close door(s) shuts and latches automatically when fusible link melts at 74°C (165°F) under fire conditions
  • Unique, concealed self-closing mechanism offers obstruction-free access to contents
  • Include polyethylene trays that sit on top of galvanized steel shelves and separate polyethylene liner for bottom sump to resist aggressive chemicals
  • Easily remove liner for quick cleaning of drips and leaks
  • Epoxy baked-on powder-coat finish, inside and out, provides increased chemical resistance
Specifications

Specifications

Blue
Self-close
2
Combi Safety Storage Cabinet
Hazardous materials
2
Videos
SDS
Documents

Documents