missing translation for 'onlineSavingsMsg'
Learn More

Justrite™ Sure-Grip™ EX Slimline Hazardous Material Safety Cabinet
GREENER_CHOICE

Catalog No. 19414807
Change view
Click to view available options
Load Bearing Capacity:
22 gal.
54 gal.
Dimensions (D x W x H):
18 x 23.25 x 65 in.
34 x 23.25 x 65 in.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Load Bearing Capacity Dimensions (D x W x H)
19414807 22 gal. 18 x 23.25 x 65 in.
19414808 54 gal. 34 x 23.25 x 65 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19414807 Supplier Justrite™ Supplier No. 862228

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Keep storage of your hazardous materials compliant with the Justrite™ Sure-Grip™ EX Hazmat Safety Cabinet.

  • Double-wall construction, dual vents, grounding wire connections, adjustable shelves
  • Leakproof sills, three-point self-latching doors and leveling feet
  • Self-close door(s) shuts and latches automatically when fusible link melts at 74°C (165°F) under fire conditions
  • Unique, concealed self-closing mechanism offers obstruction-free access to contents
  • Include polyethylene trays that sit on top of galvanized steel shelves and separate polyethylene liner for bottom sump to resist aggressive chemicals
  • Easily remove liner for quick cleaning of drips and leaks
  • Epoxy baked-on powder-coat finish, inside and out, provides increased chemical resistance

Specifications

Product Type Combi Safety Storage Cabinet
Dimensions (D x W x H) 18 x 23.25 x 65 in.
Door Style Self-close
No. of Shelves 3
No. of Doors 1
Load Bearing Capacity 22 gal.
Color Blue
For Use With (Application) Hazardous materials