missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Justrite™ Sure-Grip™ EX Compac Hazardous Material Steel Safety Cabinet

Description
- Feature all-welded, 18-gauge (1mm) double-walled steel construction with 38mm (1.5 in.) air space, 51mm (2 in.) leakproof sump
- U-Loc™ handle, dual vents with flame arresters, adjustable self-leveling feet, and ground wire connector
- Application-specific labels identify contents to safely store and segregate incompatible chemicals
- Self-closing door(s) with three-point, stainless steel bullet latching system securely close for maximum protection under fire conditions
- Self-index and self-latch to close in sequence to ensure tight closure from top to bottom
- For increased chemical resistance against leaks or drips, steel SpillSlope™ adjustable shelves include a polyethylene tray that sits on top of shelf, as well as separate polyethylene liner for bottom sump
Specifications
Specifications
| Product Type | Combi Safety Storage Cabinet |
| Dimensions (D x W x H) | 457 x 591 x 889mm |
| Door Style | Self-close |
| Load Bearing Capacity | 12 gal. |
| No. of Shelves | 1 |
| Color | Blue |
| No. of Doors | 1 |
| For Use With (Application) | Hazardous materials |