missing translation for 'onlineSavingsMsg'
Learn More

Justrite™ Sure-Grip™ EX Compac Hazardous Material Steel Safety Cabinet
GREENER_CHOICE

Catalog No. 19414803
Change view
Click to view available options
Load Bearing Capacity:
12 gal.
15 gal.
Dimensions (D x W x H):
457 x 591 x 889mm
457 x 591 x 1118mm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Load Bearing Capacity Dimensions (D x W x H)
19414803 12 gal. 457 x 591 x 889mm
19414804 15 gal. 457 x 591 x 1118mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19414803 Supplier Justrite™ Supplier No. 861228

Store hazardous materials safely even when available space is at a minimum with the Justrite™ Sure-Grip™ Safety Cabinet.

  • Feature all-welded, 18-gauge (1mm) double-walled steel construction with 38mm (1.5 in.) air space, 51mm (2 in.) leakproof sump
  • U-Loc™ handle, dual vents with flame arresters, adjustable self-leveling feet, and ground wire connector
  • Application-specific labels identify contents to safely store and segregate incompatible chemicals
  • Self-closing door(s) with three-point, stainless steel bullet latching system securely close for maximum protection under fire conditions
  • Self-index and self-latch to close in sequence to ensure tight closure from top to bottom
  • For increased chemical resistance against leaks or drips, steel SpillSlope™ adjustable shelves include a polyethylene tray that sits on top of shelf, as well as separate polyethylene liner for bottom sump

Specifications

Product Type Combi Safety Storage Cabinet
Dimensions (D x W x H) 457 x 591 x 889mm
Door Style Self-close
Load Bearing Capacity 12 gal.
No. of Shelves 1
Color Blue
No. of Doors 1
For Use With (Application) Hazardous materials