missing translation for 'onlineSavingsMsg'
Learn More

Justrite™ QuickBerm™ Rigid Lock Spill Containment System

Catalog No. 19111053
Change view
Click to view available options
Capacity (English):
1075 gal.
1435 gal.
1795 gal.
1910 gal.
2390 gal.
2990 gal.
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English)
19111053 1075 gal.
19111054 1435 gal.
19111055 1795 gal.
19111057 2390 gal.
19111056 1910 gal.
19111058 2990 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 19111053 Supplier Justrite™ Supplier No. 28522

Safe, drive-in spill containment for drums, IBC totes and tanks

Ergonomic design provides 100% inside berm accessibility with an unobstructed side-to-side floor space for vehicle tires, foot traffic and cleaning. The patented Rigid-Lock wall-support design protects personnel against trips and falls when walking along the outside perimeter. Built-in stainless steel wall supports lock the sidewalls in a 12 in. (305-mm) high, 90° vertical position. The Rigid-Lock QuickBerm™ offers immediate, secure deployment and no-worry, spill-readiness.

  • Made of UV resistant PVC coated fabric
  • Portable design folds down quickly for storage and transport
  • Single-piece construction
  • Easy setup with no assembly required
  • Includes reinforced CriticalCorner™ design featuring wraparound diagonal radio frequency welds to prevent billowing
  • Walls include 1 in. (25mm) fray-resistant finished hem along top rim for added wall strength and integrity
  • Support with brass grommet for anchoring
  • Resist extreme temperatures (-50°F to +160°F)

  • In down position, they can withstand vehicle weights in excess of 11,000 lb. (5,000kg) at each tire
  • In up position, supports will hold up to wind loads of 40miles per hour
  • Performs at full rim containment height–meaning the berm is entirely full- without bend or buckles
  • Replcaeable stainless steel support rods

Specifications

Capacity (English) 1075 gal.
Capacity (Metric) 4069 L
Material PVC-coated Fabric
Type Spill Containment System
For Use With (Equipment) Drums, IBC totes, tanks