missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Justrite™ Outdoor Safety Locker Accessory, R-11 Insulations
Description
- Installed in ceiling, walls, and sump walls for added temperature control when used in conjunction with air conditioning and heating accessories
- Must be purchased in conjunction with locker to ensure proper installation
Warranty
One year limited warranty
Specifications
Specifications
| For Use With (Equipment) | 2-drum, Fire-rated Locker Only |
| Description | R-11 Insulation |
| Color | Yellow |