missing translation for 'onlineSavingsMsg'
Learn More

Justrite™ Outdoor Safety Locker Accessory, R-11 Insulations

Catalog No. 18666629
Change view
Click to view available options
For Use With (Equipment):
12-drum, Fire-rated Locker Only
16-drum, Fire-rated Locker Only
2-drum, Fire-rated Locker Only
4-drum, Fire-rated Locker Only
6-drum, Fire-rated Locker Only
9-drum, Fire-rated Locker Only
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
18666629 2-drum, Fire-rated Locker Only
18666630 4-drum, Fire-rated Locker Only
18666631 6-drum, Fire-rated Locker Only
18666632 9-drum, Fire-rated Locker Only
18666633 12-drum, Fire-rated Locker Only
18666634 16-drum, Fire-rated Locker Only
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18666629 Supplier Justrite™ Supplier No. 915601

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For use in conjunction with heating and cooling packages to provide optimum temperature control

  • Installed in ceiling, walls, and sump walls for added temperature control when used in conjunction with air conditioning and heating accessories
  • Must be purchased in conjunction with locker to ensure proper installation

Warranty

One year limited warranty

Specifications

For Use With (Equipment) 2-drum, Fire-rated Locker Only
Description R-11 Insulation
Color Yellow