missing translation for 'onlineSavingsMsg'
Learn More

Justrite™ Mini-Berm Flex Spill Control Tray

Catalog No. 19111031
Change view
Click to view available options
Capacity (English):
30 gal.
55 gal.
70 gal.
110 gal.
Dimensions (L x W x H):
1.2 x 1.2 x 0.15 m
1.5 x 1.2 x 0.15 m
1.8 x 1.5 x 0.15 m
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English) Dimensions (L x W x H)
19111031 110 gal. 1.8 x 1.5 x 0.15 m
19111029 55 gal. 1.2 x 1.2 x 0.15 m
19111030 70 gal. 1.5 x 1.2 x 0.15 m
19111028 30 gal. 1.8 x 1.5 x 0.15 m
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19111031 Supplier Justrite™ Supplier No. 28448

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Compact, portable spill control for harsh and hazardous chemicals

  • For tabletop parts cleaning and fluid transfer operations involving petroleum products and acids
  • Made from robust modified PVC-coated fabric
  • Features rigid corner supports and sturdy foam walls
  • Includes 1 in. NPT drain fitting and utility strap
  • Fluids for disposal can be transfered by attching hose
  • Easy to fold and store

Specifications

Capacity (English) 110 gal.
Capacity (Metric) 416 L
Material PVC-coated Fabric
Type Spill Control Tray
Includes 1 in. NPT drain fitting/utility strap
Dimensions (L x W x H) 1.8 x 1.5 x 0.15 m