missing translation for 'onlineSavingsMsg'
Learn More

Justrite™ Sure-Grip™ EX Undercounter Corrosives/Acid Steel Safety Cabinet
GREENER_CHOICE

Catalog No. 17154C
Change view
Click to view available options
Weight (English):
192 lb.
194 lb.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Weight (English)
17154C 192 lb.
19033720 194 lb.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 17154C Supplier Justrite™ Supplier No. 892302

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Helps keep floor space free from hazards with a Sure-Grip EX Undercounter Cabinet. Suited for storing corrosives and acids away for added workplace safety.

Safely store corrosives under a workbench or lab counter for quick and easy access with these Justrite blue steel Undercounter safety cabinets for corrosives, which have the same quality safety features as the cabinets for flammables.

  • All models come with double-wall construction, dual vents, grounding wire connections, adjustable shelves, leakproof sills, three-point self-latching doors and leveling feet.
  • Self-close door(s) shuts and latches automatically when a fusible link melts at 165°F (74°C) under fire conditions
  • Unique, concealed self-closing mechanism offers obstruction-free access to contents
  • Polyethylene trays attached to galvanized steel shelves and a separate polyethylene liner for the bottom sump resist aggressive chemicals
  • Easy-to-remove liner offers quick cleaning of drips and leaks
  • All epoxy baked-on powder-coat finish, inside and out, provides increased chemical resistance

Compliance

Self Close models meet OSHA, NFPA 30, NFPA 1 and the International Fire Code. Cabinets are FM approved.

Warranty

10 Years

Specifications

Dimensions (D x W x H) Interior 18.75 x 31.75 x 27.37 in.
Load Bearing Capacity 22 gal.
Product Type Undercounter storage cabinet
Width (Metric) Exterior 88.9 cm
Width (Metric) Interior 80.6 cm
Depth (Metric) Exterior 68.5 cm
Height (Metric) Exterior 88.9 cm
Depth (Metric) Interior 47.6 cm
Weight (Metric) 87 kg
No. of Doors 2
Height (Metric) Interior 69.5 cm
No. of Shelves 1
For Use With (Application) Flammables
Width (English) Exterior 35 in.
Material Steel (CR)
Surface Coating Epoxy
Height (English) Exterior 35 in.
Color Blue
Door Style Manual-close
Weight (English) 192 lb.
Certifications/Compliance FM, NFPA, OSHA, U (NFPA)
Includes Double wall, dual vents, grounding wire connections, adjustable shelves, leakproof sills, three-point self latching doors and leveling feet, Polyethylene trays, separate Polyethylene liner
Depth (English) Exterior 22 in.
Depth (English) Interior 18.75 in.
Height (English) Interior 27.37 in.
Width (English) Interior 31.75 in.
Show More Show Less