Learn More
Justrite™ I'm Easy Funnel
Catalog No. 19123397
Change view
Click to view available options
Material:
Polypropylene
Catalog No. Material
19123397 Polypropylene
1 options
Catalog No. 19123397 Supplier Justrite™ Supplier No. 11202Y
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Aids pouring into smaller openings
  • Easily attaches to 1 gal. or larger Type I safety cans
  • Large rounded bowl provides protection from splashing and spills
  • Innovative hinged design allow pouring or filling from can without removing funnel
  • Durable polypropylene holds up to heavy use
Specifications
Material Polypropylene
Height (Metric) 28.6 cm