Learn More
Justrite™ Horizontal and Vertical Drum Vents
Catalog No. 17186A
Change view
Click to view available options
Includes:
Operating Valve
Operating Valve, Plated Steel Flame Arrester
Operating Valve, Stainless Steel Flame Arrester
Product Type:
Horizontal Drum Vent
Vertical Drum Vent
Material:
Brass
Non-metallic Cast Iron
Stainless Steel
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes Product Type Material
17186A Operating Valve Vertical Drum Vent Brass
17186B Operating Valve, Plated Steel Flame Arrester Vertical Drum Vent Non-metallic Cast Iron
17186D Operating Valve, Stainless Steel Flame Arrester Vertical Drum Vent Non-metallic Cast Iron
1798647 Operating Valve Horizontal Drum Vent Brass
19018564 Operating Valve Horizontal Drum Vent Stainless Steel
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 17186A Supplier Justrite™ Supplier No. 08101
For controlled release of flammable vapors; prevent explosions caused by pressure buildup
  • Choose brass, stainless steel or nonmetallic vent
  • For petroleum-based liquids, use non-metallic or brass vents
  • For chlorinated solvents, use nonmetallic vent with stainless-steel flame arrester
  • For corrosives and general solvents, use stainless steel vents
Vertical Drum Vent:
  • Has operating valve that opens automatically to prevent pressure buildup inside drum
Horizontal Drum Vent
  • Has operating valve that can be opened manually to relieve internal pressure
Order Info
All vents fit 2 in. (5.1cm) bung. Cast Iron EL Fitting for use with 17-186A and 17-186B also available; see cat. no. 19018555.
Certifications
FM approved.
Specifications
Certifications/Compliance FM
Product Type Vertical Drum Vent
Description Opens at 5 psig Internal Pressure
For Use With (Application) Controlled release of flammable vapors (Petroleum-based liquids), prevent explosions caused by pressure buildup
For Use With (Equipment) Vertical Drums
Material Brass
Includes Operating Valve