Learn More
Justrite™ Drum Siphon Adapter
Catalog No. 296357347
Change view
Click to view available options
For Use With (Equipment):
30 and 55 gal. Draining Drums
Catalog No. For Use With (Equipment)
296357347 30 and 55 gal. Draining Drums
1 options
Catalog No. 296357347 Supplier Justrite™ Supplier No. 08311
For draining 30 and 55 gallon drums
  • One end of the adapter accepts a standard 1.9cm (0.75 in.) drum faucet
  • Opposite end screws into a 1.9cm (0.75 in.) drum bung opening
  • Adjustable collar
Specifications
Product Type Drum Siphon Adapter
For Use With (Application) Empties residual solvents from a typical two to three gallons down to the last few ounces, eliminating the need for dangerous drum tipping or rolling
For Use With (Equipment) 30 and 55 gal. Draining Drums
Material Brass
Includes Adjustable Collar