Learn More
Justrite™ Drum Faucets, Extender, and Valve
Description
- Self-closing brass safety faucets feature a PTFE fluorocarbon resin seal, flame arrester and standard 0.75 in. NPT thread
- Brass faucets screw into drum; adjustable model adjusts to spout-down position
- Flexible faucet extensions provide accuracy, prevent spills; available in 8 in. long (20.3cm) brass and 6 in. long (15.2cm) stainless steel
- Brass drum gate valve fits 2 in. (5.1cm) NPT openings and has wide mouth for easy dispensing of viscous liquids from drums in horizontal position
Certifications
Faucets and valve are FM approved.
Specifications
Specifications
| Certifications/Compliance | FM |
| Product Type | Adjustable Drum Faucet |
| Material | Brass |
| Includes | PTFE Fluorocarbon Resin seal, internal flame arrester and |