missing translation for 'onlineSavingsMsg'
Learn More

Justrite™ Document Storage Box Labels

Catalog No. 191304776
Change view
Click to view available options
Color:
Yellow
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
191304776 Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 191304776 Supplier Justrite™ Supplier No. JMC23310

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For use with document storage boxes

  • Includes four yellow, two blank and two printed (SDS and instruction manual) labels

Specifications

Product Type Storage Box Label
Color Yellow
Material Polyethylene
Includes 2 blank labels and labels printed SDS and instruction manual