missing translation for 'onlineSavingsMsg'
Learn More

Justrite™ Sure-Grip™ EX Compac Corrosives/Acid Steel Safety Cabinet
GREENER_CHOICE

Catalog No. 19413822
Change view
Click to view available options
Dimensions (D x W x H) Interior:
14.56 x 20 x 30.5 in.
14.56 x 20 x 39.6 in.
Load Bearing Capacity:
12 gal. (45 L)
15 gal. (57 L)
Certifications/Compliance:
FM, NFPA, OSHA
NFPA, OSHA
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (D x W x H) Interior Load Bearing Capacity Certifications/Compliance
19413822 14.56 x 20 x 39.6 in. 15 gal. (57 L) FM, NFPA, OSHA
19159719 14.56 x 20 x 30.5 in. 12 gal. (45 L) NFPA, OSHA
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options

These cabinets are not recommended for storage of phenol, nitric acid or sulfuric acid.

Catalog No. 19413822 Supplier Justrite™ Supplier No. 891502

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

These cabinets are not recommended for storage of phenol, nitric acid or sulfuric acid.

Get more space to store hazardous material in your work environment. Justrite Sure-Grip™ EX Compac Corrosives/Acid Steel Safety Cabinet smaller sized cabinets fit nearly anywhere.

  • Flame-coated thermoplastic liner
  • 100% seamless liner on all interior walls, ceiling, sump, inside doors and shelves
  • Seam-free surface
  • Non-porous surface resists staining and cleans-up easily
  • Dual-benefits/Multi-use - complete protection from harsh acids and corrosive chemicals
  • Blue color
  • Features double-wall, 18-gauge (1mm) welded steel construction, 2 in. (51mm) liquid-tight containment sump, dual vents
  • Adjustable shelves, three-point stainless steel bullet self-latching doors with ULoc™ padlockable handle and Haz-Alert™ reflective warning labels
  • ChemCor™ liner interior

Compliance

Compliance: Retains FM Approval & Complies with OSHA regulations and the International Fire Code of NFPA 1 Uniform Fire Code. Only NFPA/OSHA for 19159712/19413871.

Specifications

Dimensions (D x W x H) Interior 14.56 x 20 x 39.6 in.
Product Type Corrosives and Acids Safety Cabinet
Height (Metric) Exterior 112 cm
Width (Metric) Interior 50.8 cm
Weight (Metric) 70 kg
Height (Metric) Interior 100.6 cm
Depth (Metric) Interior 37 cm
Width (Metric) Exterior 59.05 cm
Depth (Metric) Exterior 45.7 cm
Load Bearing Capacity 15 gal. (57 L)
No. of Doors 1
No. of Shelves 1
Certifications/Compliance FM, NFPA, OSHA
For Use With (Application) Flammables
Height (English) Exterior 44 in.
Surface Coating Epoxy
Door Style Manual-close
Depth (English) Exterior 18 in.
Weight (English) 155 lb.
Material Steel (CR)
Includes Double wall, dual vents, grounding wire connections, adjustable shelves, leakproof sills, three-point self latching doors and leveling feet, Polyethylene trays, separate Polyethylene liner
Height (English) Interior 39.6 in.
Color Blue
Width (English) Interior 20 in.
Depth (English) Interior 14.56 in.
Width (English) Exterior 23.25 in.
Show More Show Less