Learn More
Justrite™ Antistatic Dual Alligator Clips
Dual alligator clips with end connectors
Supplier:  Justrite™ 08500
Catalog No. 17-985-90E
Only null left
Explore available promotions
Description
Description
Compliance
Comply with OSHA standards and local safety codes requiring grounded containers during the transfer of liquids and drums connected to an earth ground.
Specifications
Specifications
Dual Alligator Clips
Zinc, Steel
36 in.
91.44 cm
Product Suggestions
Product Suggestions