Learn More
Beckman Coulter Beckman Coulter JS-5.3 AllSpin Swinging-Bucket Rotor, 4 x 500 mL (2,000 mL), Anodized Aluminum
Numéro de catalogue. NC1617234
Encompass
The Beckman Coulter JS-5.3 AllSpin swinging-bucket rotor (MPN 368690) is a four-place anodized-aluminum rotor and bucket assembly designed for medium-speed separations. It accepts adapters to accommodate a wide range of tubes and bottles and is suitable for rapid sedimentation of proteins, cells, and large particles, and for serum separation from whole blood.
  • Four-place swinging-bucket rotor accepting adapters to accommodate a range of tubes and bottles.
  • Anodized aluminum construction for durability and corrosion resistance.
  • Supports up to 5,300 rpm and up to 6,870 xg (for 500 mL conicals).
  • Nominal capacity of 2,000 mL (4 x 500 mL).
  • k-factor of 7,730 for sedimentation efficiency comparison.
  • Buckets interchangeable with microplate carriers.
  • Suitable for rapid sedimentation of proteins, cells, and large particles, and for serum separation from whole blood.
Numéro de catalogue. NC1617234 Fournisseur Beckman Coulter Code fournisseur 368690
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
The Beckman Coulter JS-5.3 AllSpin swinging-bucket rotor (MPN 368690) is a four-place anodized-aluminum rotor and bucket assembly designed for medium-speed separations. It accepts adapters to accommodate a wide range of tubes and bottles and is suitable for rapid sedimentation of proteins, cells, and large particles, and for serum separation from whole blood.
  • Four-place swinging-bucket rotor accepting adapters to accommodate a range of tubes and bottles.
  • Anodized aluminum construction for durability and corrosion resistance.
  • Supports up to 5,300 rpm and up to 6,870 xg (for 500 mL conicals).
  • Nominal capacity of 2,000 mL (4 x 500 mL).
  • k-factor of 7,730 for sedimentation efficiency comparison.
  • Buckets interchangeable with microplate carriers.
  • Suitable for rapid sedimentation of proteins, cells, and large particles, and for serum separation from whole blood.