Learn More
Velp™ Scientific JP Pump for Fume Aspiration
JP Recirculating Water Pump ensures fume aspiration during digestion and guarantees optimal suction.
Supplier:  Velp™ Scientific F30640198
Catalog No. 11392217
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
  • Fume aspiration depends on digestion phases and the number of samples in the digester to which the pump is connected
  • Structure is highly resistant to chemical corrosion
  • Provides considerable water saving thanks to continuous water recirculation in its tank
  • Designed to offer high performance with a flow rate of up to 35 L/min.
Specifications
Specifications
115 V
ABS
Water Pump
35 L/min.
60 Hz
Product Suggestions
Product Suggestions