Learn More
Chemglass Inc Clamp, Keck, Brown, Standard Taper, Fits Joint Size 45/50
Catalog No. CG14509
Encompass
Clamp, Keck, Brown, Standard Taper, Fits Joint Size 45/50. Made of Delrin, the clamp is non-corrosive and will not scratch glass. Can be used to 140 C. Color coded for easy identification. SOLD IN PACKAGES OF 10.
Catalog No. CG14509 Supplier Chemglass Inc Supplier No. CG 145 09
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Clamp, Keck, Brown, Standard Taper, Fits Joint Size 45/50. Made of Delrin, the clamp is non-corrosive and will not scratch glass. Can be used to 140 C. Color coded for easy identification. SOLD IN PACKAGES OF 10.