Clamp, Keck, Green, Standard Taper, Fits Joint Sizes 24/25 & 24/40., Made of Delrin�, the clamp is non-corrosive and will not scratch glass. Can be used to 140� C. Color coded for easy identification. SOLD IN PACKAGES OF 10.
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Clamp, Keck, Green, Standard Taper, Fits Joint Sizes 24/25 & 24/40., Made of Delrin�, the clamp is non-corrosive and will not scratch glass. Can be used to 140� C. Color coded for easy identification. SOLD IN PACKAGES OF 10.