missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ 96-Well, Cell Culture-Treated, U-Shaped-Bottom Microplate

Color:Clear; Sterility:Sterile; Lid:With Lid

Catalog No. FB012932


Only null left

Description

Description

  • Uniform well volume to ensure equal growth surface area
  • Well surface smooth, free from striation to maximize usable growth area
  • Raised rims on wells with uniform rings on lid to reduce evaporation
  • Single position lid reduces risks of cross-contamination, handling mistakes
  • Alphanumeric code on wells for easy identification
  • Suitable for all common instruments, automation
  • Sterilized by gamma irradiation
  • Non-pyrogenic

Warranty

Three years

Specifications

Specifications

CE
With Lid
8 x 12
Clear
Virgin Polystyrene
96
Cell Culture
Tissue culture
Yes
Standard
0.32 cm2
1/Pk., 100/Cs.
Tissue Culture Plate
0.32 mL
Sterile
Round Bottom (U-well)
100 Plates
Raised rims with condensation rings
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions