missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAPA Professional Jeresette 301 FSC Certified Latex Gloves
Offers sustainable hand protection made from FSC™ certified latex, designed for long-term use in tough work environments.
Supplier: MAPA Professional 34301028
Description
- Provides maximum comfort and durability for long-term use in demanding environments.
- Made from FSC™ certified latex, offering sustainable protection and eco-friendly performance.
- Anatomically shaped with a seamless liner to ensure freedom of movement and all-day comfort.
- High tear resistance for enhanced durability and extended glove lifespan.
- Resistant to diluted acids and bases, delivering reliable chemical protection.
- Silicone-free material prevents defects on glass and metal surfaces prior to painting.
- Individually packaged with optimized materials, reducing plastic use and incorporating 30% recycled content.
- Certified chemical protection: CAT. 3 0334 2131X Type B KPT, compliant with stringent safety standards.
- This product is not classified as hazardous under Regulation (EC) No. 1272/2008 of the European Parliament and of the Council. It contains no more than 0.1% of substances of very high concern (SVHC) and no substances listed in Annex XVII of Regulation (EC) No. 1907/2006 (REACH). UE type certificate / CE type examination certificate: 0075/014/162/05/22/0905 EX 01/05/22.
Specifications
| ANSI 2 | |
| Industrial, Laboratory | |
| 45 mil | |
| No | |
| 45 mil | |
| Chemical and Liquid Protection Gloves | |
| 45 mil | |
| EN388: 2131x | |
| B | |
| Textile Support | |
| Jersette 301 | |
| 30 cm |
| No | |
| Blue | |
| 1.15 mm | |
| Latex | |
| 1.15 mm | |
| Large | |
| 1.15 mm | |
| Type B | |
| Pebble Embossed | |
| Seamless Lining | |
| 12 in. | |
| Jersette |