missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Jenner's Stain

Catalog No. AAJ6107714
Change view
Click to view available options
Quantity:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6107714 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ6107714 Supplier Thermo Scientific Chemicals Supplier No. J6107714
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For use in staining blood smears

For use in staining blood smears

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 62851-42-7
Synonym Eosin-Methylene Blue

Specifications

Quantity 25g
Format Powder
For Use With (Application) For use in staining blood smears
Color Dark Green
Product Type Hematology Stain

RUO – Research Use Only