missing translation for 'onlineSavingsMsg'
Learn More

Janus Green B, TCI America™

Catalog No. J00025G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
J00025G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. J00025G Supplier TCI America Supplier No. J00025G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Supravital stain used for staining mitochondria in living cells.

Chemical Identifiers

CAS 2869-83-2
Molecular Formula C30H31ClN6
Molecular Weight (g/mol) 511.07
MDL Number MFCD00011758
InChI Key XXACTDWGHQXLGW-UHFFFAOYSA-M
PubChem CID 76123
ChEBI CHEBI:21184
IUPAC Name 3-(diethylamino)-7-{2-[4-(dimethylamino)phenyl]diazen-1-yl}-5-phenyl-5λ⁵-phenazin-5-ylium chloride
SMILES [Cl-].CCN(CC)C1=CC=C2N=C3C=CC(=CC3=[N+](C3=CC=CC=C3)C2=C1)N=NC1=CC=C(C=C1)N(C)C

Specifications

Color Black/Green
Quantity 5 g
Physical Form Crystal/Powder
Formula Weight 511.07
Percent Purity >= 80
Grade Research
Chemical Name or Material Janus Green B
RTECSNumber SG1633000
TSCA Yes